siRNA Details
| Gene Symbol: Olfr822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146671.1 | NP_666882.1 | 1 | 302 | ||
| GenBank mRNA | BC127972.1 | - | 1 | 306 | 40135122 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr822| Gene Symbol: Olfr822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146671.1 | NP_666882.1 | 1 | 302 | ||
| GenBank mRNA | BC127972.1 | - | 1 | 306 | 40135122 | |