siRNA Details
| Gene Symbol: Olfr402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146708.1 | NP_666919.1 | 1 | 20 | ||
| GenBank mRNA | BC116954.1 | - | 1 | 61 | 40126273 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr402| Gene Symbol: Olfr402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146708.1 | NP_666919.1 | 1 | 20 | ||
| GenBank mRNA | BC116954.1 | - | 1 | 61 | 40126273 | |