siRNA Details
| Gene Symbol: Olfr912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146810.2 | NP_667021.2 | 2 | 854 | ||
| GenBank mRNA | CB174340.1 | - | 1 | 528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr912| Gene Symbol: Olfr912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146810.2 | NP_667021.2 | 2 | 854 | ||
| GenBank mRNA | CB174340.1 | - | 1 | 528 | ||