siRNA Details
| Gene Symbol: Olfr871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146903.2 | NP_667114.2 | 1 | 686 | ||
| GenBank mRNA | BC119201.1 | - | 1 | 592 | 8733950 | |
| BC119203.1 | - | 1 | 592 | 8733952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr871| Gene Symbol: Olfr871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146903.2 | NP_667114.2 | 1 | 686 | ||
| GenBank mRNA | BC119201.1 | - | 1 | 592 | 8733950 | |
| BC119203.1 | - | 1 | 592 | 8733952 | ||