siRNA Details
Gene Symbol: Olfr871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146903.2 | NP_667114.2 | 1 | 934 | ||
GenBank mRNA | BC119201.1 | - | 1 | 840 | 8733950 | |
BC119203.1 | - | 1 | 840 | 8733952 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146903.2 | NP_667114.2 | 1 | 934 | ||
GenBank mRNA | BC119201.1 | - | 1 | 840 | 8733950 | |
BC119203.1 | - | 1 | 840 | 8733952 |