siRNA Details
| Gene Symbol: Olfr142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146984.1 | NP_667195.1 | 1 | 897 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr142| Gene Symbol: Olfr142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146984.1 | NP_667195.1 | 1 | 897 | ||