siRNA Details
| Gene Symbol: Olfr1351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147040.1 | NP_667251.1 | 1 | 524 | ||
| GenBank mRNA | BC104112.1 | - | 1 | 524 | 40043939 | |
| BC104113.1 | - | 1 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1351| Gene Symbol: Olfr1351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147040.1 | NP_667251.1 | 1 | 524 | ||
| GenBank mRNA | BC104112.1 | - | 1 | 524 | 40043939 | |
| BC104113.1 | - | 1 | 523 | |||