siRNA Details
| Gene Symbol: Olfr57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147041.2 | NP_667252.1 | 1 | 411 | ||
| GenBank mRNA | AF102534.1 | - | 1 | 144 | ||
| BC141307.1 | - | 1 | 415 | 9055919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr57| Gene Symbol: Olfr57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147041.2 | NP_667252.1 | 1 | 411 | ||
| GenBank mRNA | AF102534.1 | - | 1 | 144 | ||
| BC141307.1 | - | 1 | 415 | 9055919 | ||