siRNA Details
| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012953.1 | - | 8 | 1556 | ||
| AK168912.1 | - | 1563 | ||||
| AY253297.1 | - | 8 | 1571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc17| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012953.1 | - | 8 | 1556 | ||
| AK168912.1 | - | 1563 | ||||
| AY253297.1 | - | 8 | 1571 | |||