siRNA Details
| Gene Symbol: Mterf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013023.2 | NP_001013041.2 | 2 | 1297 | ||
| NM_172135.2 | NP_742147.1 | 2 | 1278 | |||
| GenBank mRNA | AY726770.2 | - | 2 | 1290 | ||
| BC038058.1 | - | 2 | 1270 | 4207737 | ||
| BC051251.1 | - | 2 | 1291 | 6490147 | ||

Mterf1a
Genomic Map