siRNA Details
| Gene Symbol: Fam149b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024512.2 | NP_001019683.1 | 5 | 699 | ||
| NM_001177633.1 | NP_001171104.1 | 5 | 699 | |||
| GenBank mRNA | AK031973.1 | - | 5 | 702 | ||
| AK037808.1 | - | 5 | 587 | |||
| AK077495.1 | - | 5 | 602 | |||
| AK129252.1 | - | 4 | 486 | |||
| BC025183.1 | - | 5 | 595 | 5011464 | ||
| BC058650.1 | - | 5 | 542 | |||
| BC059221.1 | - | 5 | 542 | |||
| BC069846.1 | - | 5 | 597 | 4196478 | ||
| BC080849.1 | - | 5 | 639 | 5717789 | ||

Fam149b
Genomic Map