siRNA Details
| Gene Symbol: Speer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173069.3 | NP_775092.3 | 4 | 545 | ||
| GenBank mRNA | AY151401.1 | - | 3 | 316 | ||
| BB610282.1 | - | 4 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer2| Gene Symbol: Speer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173069.3 | NP_775092.3 | 4 | 545 | ||
| GenBank mRNA | AY151401.1 | - | 3 | 316 | ||
| BB610282.1 | - | 4 | 547 | |||