siRNA Details
| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175109.3 | NP_780318.1 | 3 | 262 | ||
| GenBank mRNA | AB233218.1 | - | 3 | 258 | ||
| AK010954.1 | - | 3 | 246 | |||
| AK075974.1 | - | 3 | 243 | |||
| BC038691.1 | - | 3 | 248 | 1495043 | ||
| BC050115.1 | - | 3 | 251 | 30017673 | ||
| BC065381.1 | - | 2 | 195 | |||
| BY237968.1 | - | 3 | 262 | |||

Rps19bp1
Genomic Map