siRNA Details
| Gene Symbol: Zfp934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162911.1 | NP_001156383.1 | 4 | 1944 | ||
| NM_175252.4 | NP_780461.2 | 6 | 1812 | |||
| GenBank mRNA | AK020133.1 | - | 4 | 905 | ||
| AK030776.1 | - | 6 | 1815 | |||
| BC021319.1 | - | 1863 | ||||
| Gene Symbol: Zfp808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039239.2 | NP_001034328.2 | 4 | 2820 | ||

Zfp934
Genomic Map