siRNA Details
| Gene Symbol: Slc22a13b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079093.1 | - | 6 | 1285 | ||
| BC147282.1 | - | 6 | 1082 | 9056356 | ||
| BC147283.1 | - | 6 | 1082 | 9056343 | ||
| BY384836.1 | - | 1 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a13b-ps| Gene Symbol: Slc22a13b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079093.1 | - | 6 | 1285 | ||
| BC147282.1 | - | 6 | 1082 | 9056356 | ||
| BC147283.1 | - | 6 | 1082 | 9056343 | ||
| BY384836.1 | - | 1 | 333 | |||