siRNA Details
| Gene Symbol: Pld5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195816.1 | NP_001182745.1 | 8 | 1343 | ||
| NM_176916.4 | NP_795890.2 | 8 | 1647 | |||
| GenBank mRNA | AK136851.1 | - | 7 | 1461 | ||
| AK136888.1 | - | 7 | 1461 | |||
| AK144116.1 | - | 8 | 1636 | |||
| BC100428.1 | - | 8 | 1086 | 30247182 | ||

Pld5
Genomic Map