siRNA Details
| Gene Symbol: Gm13103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177571.3 | NP_808239.2 | 4 | 1721 | ||
| GenBank mRNA | AK054347.1 | - | 4 | 1723 | ||
| BB555205.2 | - | 1 | 520 | |||
| BC096438.1 | - | 4 | 1718 | 30481183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13103| Gene Symbol: Gm13103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177571.3 | NP_808239.2 | 4 | 1721 | ||
| GenBank mRNA | AK054347.1 | - | 4 | 1723 | ||
| BB555205.2 | - | 1 | 520 | |||
| BC096438.1 | - | 4 | 1718 | 30481183 | ||