siRNA Details
| Gene Symbol: Zfp82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253385.1 | NP_001240314.1 | 6 | 609 | ||
| NM_177889.5 | NP_808557.2 | 6 | 609 | |||
| GenBank mRNA | AK037211.1 | - | 6 | 606 | ||
| BC060530.1 | - | 5 | 536 | 5684819 | ||
| U29496.1 | - | 2 | 127 | |||

Zfp82
Genomic Map