siRNA Details
| Gene Symbol: Kiss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF472576.1 | - | 1 | 125 | ||
| BC117046.1 | - | 1 | 110 | 40126365 | ||
| BC119348.1 | - | 1 | 110 | 8734097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kiss1| Gene Symbol: Kiss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF472576.1 | - | 1 | 125 | ||
| BC117046.1 | - | 1 | 110 | 40126365 | ||
| BC119348.1 | - | 1 | 110 | 8734097 | ||