siRNA Details
| Gene Symbol: Camk2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039138.2 | NP_001034227.1 | 2 | 245 | ||
| NM_001039139.2 | NP_001034228.1 | 2 | 245 | |||
| NM_178597.5 | NP_848712.2 | 2 | 245 | |||
| GenBank mRNA | AF395884.1 | - | 2 | 122 | ||
| AK031213.1 | - | 2 | 208 | |||
| AK054406.1 | - | 2 | 212 | |||
| AK078311.1 | - | 2 | 189 | |||
| AK081969.1 | - | 2 | 207 | |||
| AK154764.1 | - | 2 | 174 | |||
| BC019162.1 | - | 2 | 197 | 3675224 | ||
| BC025597.1 | - | 2 | 199 | 5342641 | ||
| CJ151263.1 | - | 2 | 245 | |||

Camk2g
Genomic Map