siRNA Details
| Gene Symbol: Eras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181548.2 | NP_853526.1 | 2 | 1064 | ||
| GenBank mRNA | AB093573.1 | - | 2 | 1064 | ||
| AK141212.1 | - | 2 | 958 | |||
| Gene Symbol: Pcsk1n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BE986911.1 | - | 1 | 137 | ||

Eras
Genomic Map