siRNA Details
| Gene Symbol: Dsg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181682.1 | NP_859010.1 | 11 | 1711 | ||
| GenBank mRNA | AK036986.1 | - | 11 | 1819 | ||
| AY192158.1 | - | 11 | 1711 | |||
| AY315940.1 | - | 1611 | ||||
| BC137927.1 | - | 11 | 1702 | 8860946 | ||

Dsg1b
Genomic Map