siRNA Details
Gene Symbol: Defb34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183035.1 | NP_898856.1 | 2 | 133 | ||
GenBank mRNA | AJ578469.1 | - | 2 | 133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Defb34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183035.1 | NP_898856.1 | 2 | 133 | ||
GenBank mRNA | AJ578469.1 | - | 2 | 133 |