siRNA Details
Gene Symbol: 2410141K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183119.3 | NP_898942.1 | 5 | 385 | ||
GenBank mRNA | AK010804.1 | - | 387 |
Gene Symbol: Gm3844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI048080.1 | - | 295 |
