siRNA Details
Gene Symbol: Krtap13-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183189.1 | NP_899012.1 | 1 | 465 | ||
GenBank mRNA | BC027551.1 | - | 1 | 465 | 1363815 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap13-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183189.1 | NP_899012.1 | 1 | 465 | ||
GenBank mRNA | BC027551.1 | - | 1 | 465 | 1363815 |