siRNA Details
| Gene Symbol: Tma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183250.2 | NP_899073.1 | 3 | 149 | ||
| GenBank mRNA | AK002863.1 | - | 3 | 150 | ||
| AK003762.1 | - | 3 | 150 | |||
| AK160705.1 | - | 3 | 147 | |||
| AK169860.1 | - | 3 | 143 | |||
| BC034602.1 | - | 3 | 131 | |||
| BC054363.1 | - | 3 | 122 | |||
| BC090400.1 | - | 3 | 122 | |||
| BC106104.1 | - | 3 | 121 | 5029265 | ||
| BC116987.1 | - | 3 | 145 | 40126306 | ||
| BC116989.1 | - | 3 | 145 | 40126308 | ||

Tma7
Genomic Map