siRNA Details
| Gene Symbol: BC061195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC061195.1 | - | 199 | 6706856 | ||
| Gene Symbol: 1700003E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005630.1 | - | 1 | 132 | ||
| BC089468.1 | - | 1 | 133 | 6775214 | ||

BC061195
Genomic Map