siRNA Details
| Gene Symbol: Gm5128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183320.3 | NP_899143.2 | 5 | 2194 | ||
| GenBank mRNA | AK029697.1 | - | 2195 | |||
| Gene Symbol: Gm7903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242937.1 | NP_001229866.1 | 2194 | |||

Gm5128
Genomic Map