siRNA Details
| Gene Symbol: Hnf4aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC025862.1 | - | 2 | 372 | 5050923 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_197983.1 | - | 372 | Chr. - - [-] on Build | ||

Hnf4aos
Genomic Map