siRNA Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171657.1 | - | 6 | 702 | ||
| AK183758.1 | - | 2 | 83 | |||
| AK189635.1 | - | 2 | 140 | |||
| BC020179.1 | - | 6 | 683 | 4913500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emc10| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171657.1 | - | 6 | 702 | ||
| AK183758.1 | - | 2 | 83 | |||
| AK189635.1 | - | 2 | 140 | |||
| BC020179.1 | - | 6 | 683 | 4913500 | ||