siRNA Details
| Gene Symbol: Zfp273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078495.1 | - | 4 | 1063 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp273| Gene Symbol: Zfp273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078495.1 | - | 4 | 1063 | ||