siRNA Details
Gene Symbol: AF067061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199060.1 | NP_951015.1 | 816 | |||
GenBank mRNA | AF067061.1 | - | 815 | |||
AF067062.1 | - | 816 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AF067061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199060.1 | NP_951015.1 | 816 | |||
GenBank mRNA | AF067061.1 | - | 815 | |||
AF067062.1 | - | 816 |