siRNA Details
| Gene Symbol: Trim30d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167828.1 | NP_001161300.1 | 4, 5 | 537 | ||
| NM_199146.2 | NP_954597.2 | 5, 6 | 1013 | |||
| GenBank mRNA | AK136456.1 | - | 4, 5 | 541 | ||
| AK164938.1 | - | 4, 5 | 536 | |||
| BC056631.1 | - | 5, 6 | 989 | 5035734 | ||

Trim30d
Genomic Map