siRNA Details
| Gene Symbol: Ugt1a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201410.3 | NP_958812.3 | 2 | 409 | ||
| GenBank mRNA | AY227198.1 | - | 1 | 365 | ||
| BC026561.1 | - | 2 | 389 | 5050488 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt1a6b| Gene Symbol: Ugt1a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201410.3 | NP_958812.3 | 2 | 409 | ||
| GenBank mRNA | AY227198.1 | - | 1 | 365 | ||
| BC026561.1 | - | 2 | 389 | 5050488 | ||