siRNA Details
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 5, 6 | 337 | ||
GenBank mRNA | BC049626.1 | - | 5, 6 | 337 | 6705850 |
Gene Symbol: 4921509O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014857.1 | - | 6, 7 | 587 |
Gene Symbol: Gm20858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AV269226.1 | - | 19 |
