siRNA Details
| Gene Symbol: Olfr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206822.1 | NP_996558.1 | 1 | 308 | ||
| GenBank mRNA | BC148303.2 | - | 1 | 308 | 40041401 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr10| Gene Symbol: Olfr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206822.1 | NP_996558.1 | 1 | 308 | ||
| GenBank mRNA | BC148303.2 | - | 1 | 308 | 40041401 | |