siRNA Details
| Gene Symbol: Olfr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206823.1 | NP_996559.1 | 1 | 305 | ||
| GenBank mRNA | BC147769.1 | - | 1 | 544 | 9056708 | |
| BC147777.1 | - | 1 | 544 | 9056711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr153| Gene Symbol: Olfr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206823.1 | NP_996559.1 | 1 | 305 | ||
| GenBank mRNA | BC147769.1 | - | 1 | 544 | 9056708 | |
| BC147777.1 | - | 1 | 544 | 9056711 | ||