siRNA Details
| Gene Symbol: Olfr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207201.1 | NP_997084.1 | 1 | 336 | ||
| Gene Symbol: Olfr1354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199840.1 | NP_001186769.1 | 1 | 336 | ||
| GenBank mRNA | CB174062.1 | - | 545 | |||

Olfr8
Genomic Map