siRNA Details
Gene Symbol: Olfr1283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207236.1 | NP_997119.1 | 1 | 481 | ||
GenBank mRNA | BC125536.1 | - | 1 | 524 | 40130051 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207236.1 | NP_997119.1 | 1 | 481 | ||
GenBank mRNA | BC125536.1 | - | 1 | 524 | 40130051 |