siRNA Details
| Gene Symbol: Vmn1r181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207546.2 | NP_997429.2 | 1 | 397 | ||
| GenBank mRNA | BC120650.1 | - | 1 | 301 | 40129573 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r181| Gene Symbol: Vmn1r181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207546.2 | NP_997429.2 | 1 | 397 | ||
| GenBank mRNA | BC120650.1 | - | 1 | 301 | 40129573 | |