siRNA Details
| Gene Symbol: Olfr1252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207568.1 | NP_997451.1 | 1 | 537 | ||
| GenBank mRNA | BC125497.1 | - | 1 | 634 | 40130012 | |
| BC125499.1 | - | 1 | 634 | 40130014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1252| Gene Symbol: Olfr1252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207568.1 | NP_997451.1 | 1 | 537 | ||
| GenBank mRNA | BC125497.1 | - | 1 | 634 | 40130012 | |
| BC125499.1 | - | 1 | 634 | 40130014 | ||