siRNA Details
| Gene Symbol: Olfr1118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC120792.1 | - | 1 | 169 | 40129715 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1118| Gene Symbol: Olfr1118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC120792.1 | - | 1 | 169 | 40129715 | |