siRNA Details
| Gene Symbol: Krt78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212487.4 | NP_997652.4 | 6, 7 | 1087 | ||
| GenBank mRNA | AK146611.1 | - | 6, 7 | 1084 | ||
| AY651827.1 | - | 6, 7 | 912 | |||
| BC119600.1 | - | 6, 7 | 1024 | |||
| BC119601.1 | - | 6, 7 | 1024 | |||
| BC127241.1 | - | 6, 7 | 1024 | |||
| BC127618.1 | - | 6, 7 | 1024 | |||

Krt78
Genomic Map