siRNA Details
Gene Symbol: KIF20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005733.2 | NP_005724.1 | 19 | 3031 | ||
GenBank mRNA | AF070672.1 | - | 18 | 2562 | ||
AF153329.1 | - | 19 | 2569 | |||
AK024804.1 | - | 8 | 1088 | |||
AK025790.1 | - | 19 | 2590 | |||
AK296879.1 | - | 20 | 2545 | |||
BC012999.2 | - | 19 | 2569 | 3604291 |
