siRNA Details
| Gene Symbol: ARHGAP39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308207.1 | NP_001295136.1 | 12 | 4689 | ||
| NM_001308208.1 | NP_001295137.1 | 11 | 4342 | |||
| NM_025251.2 | NP_079527.1 | 11 | 4308 | |||
| XM_011517308.1 | XP_011515610.1 | 12 | 4589 | |||
| XM_011517309.1 | XP_011515611.1 | 12 | 4493 | |||
| XM_011517312.2 | XP_011515614.1 | 10 | 5127 | |||
| XM_017013870.1 | XP_016869359.1 | 13 | 5661 | |||
| XM_017013871.1 | XP_016869360.1 | 12 | 4720 | |||
| GenBank mRNA | AB051475.1 | - | 12 | 4693 | ||
| AK092501.1 | - | 7 | 1755 | |||
| BC013071.2 | - | 5 | 1619 | |||

ARHGAP39
Genomic Map