siRNA Details
| Gene Symbol: Ddit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290183.1 | NP_001277112.1 | 3 | 437 | ||
| NM_007837.4 | NP_031863.3 | 4 | 485 | |||
| GenBank mRNA | AK028120.1 | - | 4 | 482 | ||
| BC013718.1 | - | 4 | 480 | 4012433 | ||
| CF982562.1 | - | 1 | 135 | |||
| X67083.1 | - | 3 | 392 | |||

Ddit3
Genomic Map