siRNA Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304363.1 | NP_001291292.1 | 2 | 226 | ||
| NM_001304364.1 | NP_001291293.1 | 2 | 355 | |||
| NM_020924.3 | NP_065975.1 | 2 | 268 | |||
| GenBank mRNA | AB046792.1 | - | 1 | 1386 | ||
| AF323460.2 | - | 1 | 153 | |||
| AK057445.1 | - | 2 | 355 | |||
| BC018748.1 | - | 2 | 217 | 4553653 | ||
| DA212731.1 | - | 2 | 226 | |||
| HY066289.1 | - | 2 | 269 | |||

ZBTB26
Genomic Map