siRNA Details
| Gene Symbol: BLVRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000713.2 | NP_000704.1 | 5 | 831 | ||
| GenBank mRNA | BC109371.1 | - | 5 | 748 | 5229026 | |
| BU528444.1 | - | 3 | 426 | |||
| D26308.1 | - | 5 | 779 | |||
| D32143.1 | - | 5 | 809 | |||
| EU794593.1 | - | 5 | 831 | |||

BLVRB
Genomic Map