siRNA Details
| Gene Symbol: KCTD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002914.2 | NP_001002914.1 | 1 | 2135 | ||
| GenBank mRNA | AK056227.1 | - | 1 | 1862 | ||
| AY646650.1 | - | 1 | 1862 | |||
| BC019929.1 | - | 1 | 1386 | 5115130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD11| Gene Symbol: KCTD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002914.2 | NP_001002914.1 | 1 | 2135 | ||
| GenBank mRNA | AK056227.1 | - | 1 | 1862 | ||
| AY646650.1 | - | 1 | 1862 | |||
| BC019929.1 | - | 1 | 1386 | 5115130 | ||