siRNA Details
| Gene Symbol: ZNF311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010877.3 | NP_001010877.2 | 3, 4 | 302 | ||
| XM_011514512.1 | XP_011512814.1 | 4, 5 | 1820 | |||
| XM_017010758.1 | XP_016866247.1 | 4, 5 | 1745 | |||
| XM_017010759.1 | XP_016866248.1 | 5, 6 | 654 | |||
| GenBank mRNA | AK289881.1 | - | 4, 5 | 480 | ||
| BC136745.1 | - | 3, 4 | 363 | 9020735 | ||

ZNF311
Genomic Map